Orders LinkContactUsLinkComponent
Goat serum
Goat serum
Goat serum, Sterile filtered, Frossen
Katalog # S2000-100
Leverandør:   Biowest
Goat serum
Katalog # S2000-100
Leverandør:  Biowest
Goat serum
Goat serum
Goat serum, Sterile filtered, Frossen
Price
236,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Goat serum is a blood derived product collected from healthy non immunised animals.
  • Blocking agent or negative control in immunoassays
  • For immunohistochemistry and immunocytochemistry, used between 1 to 20% as a blocking agent in various studies
Suitable alternative to FBS in a wide range of cell culture applications (e.g. isolation and identification of viruses produced in a variety of primary cells).
Specifikationer
  • Serum format:
    Frossen
  • Sterilitet:
    Sterile filtered
  • Beskrivelse:
    Goat serum
  • Seq:
    01