Replacement Erlenmeyer flask caps, Corning®
Ventileret låg, 70 mm, til 3 L kolbe
Leverandør: Corning
Gå til:
Product Details & Documents
Made of PP.
Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.
Advarsel: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifikationer
- Beskrivelse:Ventileret låg, 70 mm, til 3 L kolbe
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



