96-well plates, polypropylene, without lids
Storage plate, 96-well, expanded volume, natural, V-bottom well, 450 µl, +, 10/bag
Leverandør: Corning
96-well plates, polypropylene, without lids
Katalog # 732-0485
Leverandør: Corning
Gå til:
Product Details & Documents
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
Specifikationer
- Pakket:10/bag
- Steril:+
- Beskrivelse:Storage plate, 96-well, expanded volume, natural, V-bottom well
- Seq:09
- Brøndvolumen:450 µl
- Cat. No.:732-0485
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Lav fremstillingspåvirkning
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



