Orders LinkContactUsLinkComponent
Screw cap for reagent bottles
SVL 22
  BIBB4503/22
 :   SciLabware
Screw cap for reagent bottles
  BIBB4503/22
Screw cap for reagent bottles
Screw cap for reagent bottles
SVL 22
575,00 kr
1 * 20 Items
Phenolic cap with PTFE faced sealing disc
  • Fits PYREX® reagent bottles
  • Excellent chemical resistance
  • Autoclavable up to 121 °C
  • Gevind:
    SVL 22