Orders LinkContactUsLinkComponent
Screw cap for reagent bottles
Screw cap for reagent bottles
SVL18
Katalog # BIBB4503/18
Leverandør:   SciLabware
Screw cap for reagent bottles
Katalog # BIBB4503/18
Leverandør:  SciLabware
Screw cap for reagent bottles
Screw cap for reagent bottles
SVL18
Price
502,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents
Phenolic cap with PTFE faced sealing disc
  • Fits PYREX® reagent bottles
  • Excellent chemical resistance
  • Autoclavable up to 121 °C
Specifikationer
  • Gevind:
    SVL18