Screw cap for reagent bottles
SVL18
Katalog # BIBB4503/18
Leverandør: SciLabware
Gå til:
Product Details & Documents
Phenolic cap with PTFE faced sealing disc
- Fits PYREX® reagent bottles
- Excellent chemical resistance
- Autoclavable up to 121 °C
Specifikationer
- Gevind:SVL18