Orders LinkContactUsLinkComponent
Screw cap for reagent bottles
Screw cap for reagent bottles
SVL 13
Catalog # BIBB4503/13
Supplier:   SciLabware
Screw cap for reagent bottles
Catalog # BIBB4503/13
Supplier:  SciLabware
Screw cap for reagent bottles
Screw cap for reagent bottles
SVL 13
Price
410,00 kr
Log in for availability
1 * 20 Items
Product Details & Documents
Phenolic cap with PTFE faced sealing disc
  • Fits PYREX® reagent bottles
  • Excellent chemical resistance
  • Autoclavable up to 121 °C
Specifications
  • Gevind:
    SVL 13