Screw cap for reagent bottles
SVL 13
Catalog # BIBB4503/13
Supplier: SciLabware
Jump to:
Product Details & Documents
Phenolic cap with PTFE faced sealing disc
- Fits PYREX® reagent bottles
- Excellent chemical resistance
- Autoclavable up to 121 °C
Specifications
- Gevind:SVL 13