Molecular models of elements, Molymod®
Halogen, grøn, 1, 20 mm
Catalog # SPIRMA-101
Marketsource
Supplier: SPIRING ENTERPRISES
Molecular models of elements, Molymod®
Catalog # SPIRMA-101
Supplier: SPIRING ENTERPRISES
Marketsource
Jump to:
Product Details & Documents
Molymod is the original, unique, dual-scale system of high quality molecular models.
Specifications
- Farve:grøn
- Beskrivelse:Halogen
- Antal huller:1
- Ø:20 mm