Centrifugerør
15 ml, ∼17×115 mm, uden cap, ungraduated
HIRS6000101
Glass.
- Graduated or ungraduated
- Good chemical resistance
Long conical or short conical form.
- Lukkestørrelse:uden cap
- Ø×L:∼17×115 mm
- Model:ungraduated
- Kapacitet:15 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



