Organic reference standards, sumatriptans, Ph. Eur., RS
Sumatriptan impurity mixture, Reference standard, 3 mg
Catalog # EDQMY0000026
Supplier: EDQM
Organic reference standards, sumatriptans, Ph. Eur., RS
Catalog # EDQMY0000026
Supplier: EDQM
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pakn:3 mg
- Standard:Reference standard
- Beskrivelse:Sumatriptan impurity mixture
- Cat. No.:EDQMY0000026
- Beskrivelse:Sumatriptan impurity mixture
- CAS-nr:103628-48-4
- Formel:C₁₈H₂₇N₃O₆S



