Organic reference standards, sumatriptans, Ph. Eur., RS
Sumatriptan succinate, Reference standard, 50 mg
Katalog # EDQMY0000022
Leverandør: EDQM
Organic reference standards, sumatriptans, Ph. Eur., RS
Katalog # EDQMY0000022
Leverandør: EDQM
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:50 mg
- Standard:Reference standard
- Beskrivelse:Sumatriptan succinate
- Cat. No.:EDQMY0000022
- Beskrivelse:Sumatriptan succinate
- CAS-nr:103628-48-4
- Formel:C₁₈H₂₇N₃O₆S
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



