Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
Cover glasses, square, without cords and blisters, 24×32 mm, #1
Katalog # KNITVD12432Y100A
Cover glasses
Katalog # KNITVD12432Y100A
Leverandør:  
Cover glasses
Cover glasses, square, without cords and blisters, 24×32 mm, #1
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
COVER GLASS 24X32MM, NR.1 1 * 1.000 Items Katalog #631-1572
Produktet 631-1572 ligner KNITVD12432Y100A, men producenten og specifikationerne kan variere.
Product Details & Documents
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certifications: ISO 8255/1
Emballering: 100 pieces per box - 1000 pieces per carton.
Documents
Specifikationer
  • Antal:
    #1
  • Beskrivelse:
    Cover glasses, square, without cords and blisters
  • L×B:
    24×32 mm