Fork clamps for spherical joints
51
Katalog # LENZ01235130
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
Clips for spherical joints made from stainless steel chrome/nickel.
- Easy to install and remove
- Excellent heat and chemical resistance
- With set screw
Specifikationer
- Til NS:51
- Seq:04