Orders LinkContactUsLinkComponent
Fork clamps for spherical joints
51
Katalog # LENZ01235130
Leverandør:   Lenz Laborglas
Fork clamps for spherical joints
Katalog # LENZ01235130
Leverandør:  Lenz Laborglas
Fork clamps for spherical joints
Fork clamps for spherical joints
51
Price
776,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Clips for spherical joints made from stainless steel chrome/nickel.
  • Easy to install and remove
  • Excellent heat and chemical resistance
  • With set screw
Specifikationer
  • Til NS:
    51
  • Seq:
    04