Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Stivelse (from potato)

Stivelse (from potato)

5 kg
Katalog # ICNA0210295405
Leverandør:   MP Biomedicals
Stivelse (from potato)
Katalog # ICNA0210295405
Leverandør:  MP Biomedicals
CAS nummer:  9005-25-8
Stivelse (from potato)

5 kg

CAS nummer:  9005-25-8
Price
1.800,00 kr
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
  • CAS nummer:9005-25-8
  • Formel:(C₆H₁₀O₅)n
  • MDL nummer:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-679-6

Documents

Specifikationer

Specifikationer

  • Pakn:
    5 kg

Specifikationer