Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Stivelse (from potato)

Stivelse (from potato)

Glass bottle for solids, 2 kg
Katalog # ICNA0210295402
Leverandør:   MP Biomedicals
Stivelse (from potato)
Katalog # ICNA0210295402
Leverandør:  MP Biomedicals
CAS nummer:  9005-25-8
Stivelse (from potato)

Glass bottle for solids, 2 kg

CAS nummer:  9005-25-8
Price
776,00 kr
Log ind for tilgængelighed
1 * 2 kg
Product Details & Documents
  • CAS nummer:9005-25-8
  • Formel:(C₆H₁₀O₅)n
  • MDL nummer:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-679-6

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2 kg

Specifikationer