Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lakmus

Lakmus

Glass bottle for solids, 25 g
Katalog # ICNA0215526325
Leverandør:   MP Biomedicals
Lakmus
Katalog # ICNA0215526325
Leverandør:  MP Biomedicals
CAS nummer:  1393-92-6
Lakmus

Glass bottle for solids, 25 g

CAS nummer:  1393-92-6
Price
1.180,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
  • CAS nummer:1393-92-6
  • MDL nummer:MFCD00131528
  • EINECS:215-739-6

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.