Capillaries for LC systems
Capillaries, PEEK coated fused silica, C/D, 650 mm, I.Ø 1905 mm, to connect injection valve to injector seat or 2nd pump
HEWLG1375-87327
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Capillaries, PEEK coated fused silica, C/D, 650 mm, I.Ø 1905 mm, to connect injection valve to injector seat or 2nd pump
- Seq:120