Capillaries for LC systems
Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
HEWLG1375-87324
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Capillaries, PEEK coated fused silica, C/D, 700 mm, I.Ø 635 mm, to connect switching valve to column
- Seq:118