Capillaries for LC systems
Capillaries, PEEK coated fused silica, C/D, 550 mm, I.Ø 635 mm, to connect injection valve/switching valve to flow sensor/column
Katalog # HEWLG1375-87323
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, PEEK coated fused silica, C/D, 550 mm, I.Ø 635 mm, to connect injection valve/switching valve to flow sensor/column
- Seq:117