Orders LinkContactUsLinkComponent
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Frit adapter, PTFE
Katalog # HEWLG1361-23205
Leverandør:   AGILENT HEWLETT PACKARD
CrossLab Bottle Head Assemblies for HPLC
Katalog # HEWLG1361-23205
CrossLab Bottle Head Assemblies for HPLC
CrossLab Bottle Head Assemblies for HPLC
Frit adapter, PTFE
Price
217,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
All the parts required to connect solvent bottles to your HPLC instrument.
  • Instrument specific ready-to-use bottle head assemblies available
  • Special ultraclean tubing for MS applications available
Agilent bottle head assemblies contain all the parts that are required to connect solvent bottles to your HPLC instrument. Each assembly contains a solvent inlet filter, tubing, a bottle cap, and fittings.
Specifikationer
  • Beskrivelse:
    Frit adapter, PTFE
  • Seq:
    11