Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Fused silica/PEEK capillary, 50 μm ID, 12 cm long, outlet
HEWLG1315-87328
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
HEWLG1315-87328
- Beskrivelse:Fused silica/PEEK capillary, 50 μm ID, 12 cm long, outlet
- Seq:23