Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Fused silica/PEEK capillary, 50 μm ID, 40 cm long, inlet
Katalog # HEWLG1315-87323
Leverandør:   AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-87323
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)

Fused silica/PEEK capillary, 50 μm ID, 40 cm long, inlet

Price
1.390,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Fused silica/PEEK capillary, 50 μm ID, 40 cm long, inlet
  • Seq:
    22
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.