Orders LinkContactUsLinkComponent
Headspace screw top vials
Headspace screw top vials
20,00 ml, 23×75 mm, klar, No
Katalog # HEWL5188-2753
Leverandør:   AGILENT HEWLETT PACKARD
Headspace screw top vials
Katalog # HEWL5188-2753
Headspace screw top vials
Headspace screw top vials
20,00 ml, 23×75 mm, klar, No
Price
694,00 kr
Log ind for tilgængelighed
1 * 100 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
The CombiPAL precision-threaded headspace screw top vials are ideal for applications in the environmental, food and beverage, industrial hygiene, drug analysis, and clinical chemistry industries.
Specifikationer
  • Kasket inkluderet:
    No
  • Diameter×H:
    23×75 mm
  • Kapacitet:
    20,00 ml
  • Farve:
    klar