Solvent filter/degasser
PTFE seal for HEWL3150-0577, PTFE
Katalog # HEWL5188-2745
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
- Degasses eluents as particulates are removed
- Increases solvent inlet lifetime
- Eliminates pump downtime caused by air locks and particulates in check valves
- Decreases piston wear, while increasing column life
Specifikationer
- Beskrivelse:PTFE seal for HEWL3150-0577
- Materiale:PTFE
- Seq:05