Orders LinkContactUsLinkComponent
Capillaries for LC systems
Flexible capillaries, red, stainless steel, without fittings, 2,3 μl, 200 mm, I.Ø 0,12 mm
Katalog # HEWL5065-9935
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWL5065-9935
Capillaries for LC systems
Capillaries for LC systems
Flexible capillaries, red, stainless steel, without fittings, 2,3 μl, 200 mm, I.Ø 0,12 mm
Price
351,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Flexible capillaries, red, stainless steel, without fittings, 2,3 μl, 200 mm, I.Ø 0,12 mm
  • Seq:
    102