Capillaries for LC systems
Flexible capillaries, stainless steel, without fittings, 20,6 μl, 105 mm, I.Ø 0,5 mm
Katalog # HEWL5065-9927
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Flexible capillaries, stainless steel, without fittings, 20,6 μl, 105 mm, I.Ø 0,5 mm
- Seq:105