Orders LinkContactUsLinkComponent
Capillaries for LC systems
Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
  HEWL5022-6510
Capillaries for LC systems
  HEWL5022-6510
Capillaries for LC systems
Capillaries for LC systems
Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
351,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
  • Seq:
    107