Capillaries for LC systems
Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
HEWL5022-6510
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Flexible capillaries, stainless steel, without fittings, 55 μl, 280 mm, I.Ø 0,5 mm
- Seq:107