Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
X-Gal enzyme substrate

X-Gal enzyme substrate

1 KIT
Katalog # R0402
Leverandør:   Thermo Fisher Scientific
X-Gal enzyme substrate
Katalog # R0402
CAS nummer:  7240-90-6
X-Gal enzyme substrate
X-Gal enzyme substrate

1 KIT

CAS nummer:  7240-90-6
Price
1.360,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Thermo Scientific X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate

Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.

Synonymer:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS nummer:7240-90-6
  • Formel:C₁₄H₁₅BrClNO₆
  • MDL nummer:MFCD00005666
  • Molecular Weight:408.63 g/mol
  • Smeltende Pt:237…239 °C
  • Opbevaringstemperatur:Fryser
  • EINECS:230-640-8
Specifikationer

Specifikationer

  • Pakn:
    1 KIT

Specifikationer