Orders LinkContactUsLinkComponent
Oligo(dT)₁₈ primer
Oligo(dT)₁₈ primer, 120 µl/60 µg (100 µM), 1 KIT
Katalog # SO132
Leverandør:   Thermo Fisher Scientific
Oligo(dT)₁₈ primer
Katalog # SO132
CAS nummer:  
Oligo(dT)₁₈ primer
Oligo(dT)₁₈ primer
Oligo(dT)₁₈ primer, 120 µl/60 µg (100 µM), 1 KIT
CAS nummer:  
Price
831,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Oligo(dT)₁₈ primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Oligo(dT)₁₈ primer has been functionally tested in first strand cDNA synthesis.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Oligo(dT)₁₈ primer, 120 µl/60 µg (100 µM)
Specifikationer