Guanidiniumchlorid
Glass bottle for solids, 250 g
Catalog # SERA24205.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Guanidinhydrochlorid
- CAS Number:50-01-1
- Formula:NH₂C(=NH)NH₂·HCl
- MDL Number:MFCD00013026
- Molecular Weight:95.53 g/mol
- Merck Index:13,04578
- Melting Pt:185 °C
- Density:1,354 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-002-3
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:250 g



