Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumdodecylsulfat (SDS)

Natriumdodecylsulfat (SDS)

Glass bottle for solids, 250 g
Katalog # SERA20765.02
Brandfarlig
Giftig
Marketsource
Leverandør:   Serva
Natriumdodecylsulfat (SDS)
Katalog # SERA20765.02
Leverandør:  Serva
Marketsource
CAS nummer:  151-21-3
Natriumdodecylsulfat (SDS)
Natriumdodecylsulfat (SDS)

Glass bottle for solids, 250 g

CAS nummer:  151-21-3
Price
373,38 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
  • CAS nummer:151-21-3
  • Formel:CH₃(CH₂)₁₁OSO₃Na
  • MDL nummer:MFCD00036175
  • Molecular Weight:288.38 g/mol
  • Merck Index:14,08636
  • Smeltende Pt:204…207 °C
  • Tæthed:1,1 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-788-1
  • FN:1325
  • ADR:4.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.