Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumdodecylsulfat (SDS)

Natriumdodecylsulfat (SDS)

Glass bottle for solids, 100 g
Catalog # SERA20765.01
Flammable
Toxic
Marketsource
Supplier:   Serva
Natriumdodecylsulfat (SDS)
Catalog # SERA20765.01
Supplier:  Serva
Marketsource
CAS Number:  151-21-3
Natriumdodecylsulfat (SDS)
Natriumdodecylsulfat (SDS)

Glass bottle for solids, 100 g

CAS Number:  151-21-3
Price
201,63 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
  • CAS Number:151-21-3
  • Formula:CH₃(CH₂)₁₁OSO₃Na
  • MDL Number:MFCD00036175
  • Molecular Weight:288.38 g/mol
  • Merck Index:14,08636
  • Melting Pt:204…207 °C
  • Density:1,1 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Stue temperatur
  • EINECS:205-788-1
  • UN:1325
  • ADR:4.1,II
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.