X-Gal
Glass bottle for solids, 250 mg
Katalog # SERA15243.06
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
- CAS nummer:7240-90-6
- Formel:C₁₄H₁₅BrClNO₆
- MDL nummer:MFCD00005666
- Molecular Weight:408.63 g/mol
- Smeltende Pt:237…239 °C
- Opbevaringstemperatur:Fryser
- EINECS:230-640-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 mg



