Agarose
Glass bottle for solids, 100 g
Katalog # SERA11384.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
- CAS nummer:9012-36-6
- MDL nummer:MFCD00081294
- Smeltende Pt:60…90 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-731-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g