Base materials
Peptone plus from meat (enzymatic digest), Glass bottle for solids, 1 kg
Katalog # SERA48605.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
- Suitable as nutrient for laboratory media and industrial fermentation
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg
- Beskrivelse:Peptone plus from meat (enzymatic digest)