Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone plus from meat (enzymatic digest), Glass bottle for solids, 1 kg
Katalog # SERA48605.02
Marketsource
Leverandør:   Serva
Base materials
Katalog # SERA48605.02
Leverandør:  Serva
Marketsource
CAS nummer:  
Base materials
Peptone plus from meat (enzymatic digest), Glass bottle for solids, 1 kg
CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg
  • Beskrivelse:
    Peptone plus from meat (enzymatic digest)
Specifikationer