Spermidin trihydrochlorid
Glass bottle for solids, 5 g
SERA39786.02
Marketsource
: Serva
: 4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride
- :334-50-9
- :C₇H₂₂Cl₃N₃
- :MFCD00012918
- :254.63 g/mol
- :12,08893
- :256…258 °C
- :Fryser
- :206-379-0
- Pakning:Glass bottle for solids
- Pakn:5 g