Ficoll® 400
Ficoll™ 400, molecular biology grade
Katalog # SERA39763.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Ficoll® 400 is a neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution. Concentrations up to 50% (w/v), covering a density range up to 1,2 g/ml, can be obtained without exceeding normal osmolarity. Ficoll™ 400 is suitable for the preparation of continuous and discontinuous density gradients, for stabilisation of membrane-bound particles, and for isolation of lymphocytes.
- Better osmotic properties than sucrose, preserving functional and morphological integrity
- Does not penetrate biological membranes
- Readily soluble in aqueous solution
- Molecular biology grade (free from DNase and RNase) is suitable for use in hybridisation buffers like Denhardt's Solution
Ficoll® is a registered trademark of Pharmacia Biotech AB.
Specifikationer
- Beskrivelse:Ficoll™ 400, molecular biology grade
- Seq:01