MES monohydrat
Glass bottle for solids, 250 g
SERA29830.02
Marketsource
: Serva
: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
- :145224-94-8
- :C₆H₁₃NO₄S·1H₂O
- :MFCD00149409
- :213.25 g/mol
- :13,05929
- :> 300 °C
- :Stue temperatur
- :224-632-3
- Pakning:Glass bottle for solids
- Pakn:250 g