Single - component oxygenates solutions
Methanol, with 10000 µg/ml in DI water, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 mL
Katalog # REST30467
Leverandør: Restek
Gå til:
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifikationer
Specifikationer
- Pakn:1 mL
- Standard:US EPA 502.2, 8021, 8206B
- Beskrivelse:Methanol, with 10000 µg/ml in DI water
- Pk Info:1 ml/ampoule
- Cat. No.:REST30467
- Beskrivelse:Methanol
- CAS-nr:67-56-1
- Formel:CH₃OH
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



