Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Columns, MXT®-5

4,5" standard coil, Kapillær, MXT®-5, 0,50 µm, 0,25 mm, 60 m
Catalog # REST70241
Supplier:   Restek
Columns, MXT®-5
Catalog # REST70241
Supplier:  Restek
Columns, MXT®-5
Columns, MXT®-5

4,5" standard coil, Kapillær, MXT®-5, 0,50 µm, 0,25 mm, 60 m

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter

General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.

Specifications
  • Kolonne form:
    Kapillær
  • Filmtykkelse:
    0,50 µm
  • Ø indv.:
    0,25 mm
  • Beskrivelse:
    4,5" standard coil
  • Længde:
    60 m
  • Seq:
    23
  • Fase:
    MXT®-5