VWR® Rectangular Cover Glasses
22×32 mm, #1,5
Catalog # 631-0134
Supplier: VWR Collection
Jump to:
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: Pakket i dispenseræsker af plast med hængslet låg.
Specifications
- Antal:#1,5
- L×B:22×32 mm
- Seq:04