Orders LinkContactUsLinkComponent
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
20×20 mm, #1
Katalog # 631-0122
Leverandør:   VWR Collection
VWR® Rectangular Cover Glasses
Katalog # 631-0122
Leverandør:  VWR Collection
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
20×20 mm, #1
Price
254,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Emballering: Pakket i dispenseræsker af plast med hængslet låg.
Specifikationer
  • Antal:
    #1
  • L×B:
    20×20 mm
  • Seq:
    02