Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumacetat 7.5 M, clear solution

Ammoniumacetat 7.5 M, clear solution

Glass bottle, 1 L
Katalog # ICNA0219875991
Irriterende
Leverandør:   MP Biomedicals
Ammoniumacetat 7.5 M, clear solution
Katalog # ICNA0219875991
Leverandør:  MP Biomedicals
CAS nummer:  631-61-8
Ammoniumacetat 7.5 M, clear solution
Ammoniumacetat 7.5 M, clear solution

Glass bottle, 1 L

CAS nummer:  631-61-8
Price
2.010,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Ammonium acetate is a chemical compound which is derived from the reaction of ammonia and acetic acid. Ammonium acetate is volatile at low pressures. Because of which it has been used to replace cell buffers with non-volatile salts, in preparing samples for mass spectrometry. It is used as a buffer for mobile phases for HPLC with ELSD detection.

Ammonium acetate is a widely used reagent in molecular biology and chromatography. Its applications include the purification and precipitation of DNA and protein crystallization. Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides. Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.

Store at +4 °C.

Presentation: Clear Solution

Synonymer:  Acetic acid ammonium salt
  • CAS nummer:631-61-8
  • Formel:H₃CCOONH₄
  • MDL nummer:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Merck Index:13,00495
  • Smeltende Pt:114 °C
  • Tæthed:1,17 (lit.)
  • Flash Pt:136 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:211-162-9
Specifikationer

Specifikationstestresultater

  • Identity Test:
    Passes
  • Concentration:
    7.5 M aq (578.25 g/L)
  • Sterility:
    Sterile

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.