Orders LinkContactUsLinkComponent
Penetration probes, K-Type
Spring-loaded V4A tube Ø 3 mm with slim insertion tip, with plastic handle, anti-buckling glanding,1 m highly flexible silicone cable and 3,5 mm gold plated jack connector
Response time (T90): water 0,4 m/s < 2 s, air 2 m/s ~40 s, −70…+200 °C
  GRSG600431
 :   Greisinger
Penetration probes, K-Type
  GRSG600431
Penetration probes, K-Type
Penetration probes, K-Type
Spring-loaded V4A tube Ø 3 mm with slim insertion tip, with plastic handle, anti-buckling glanding,1 m highly flexible silicone cable and 3,5 mm gold plated jack connector
Response time (T90): water 0,4 m/s < 2 s, air 2 m/s ~40 s, −70…+200 °C
1 * 1 Items
  • Område:
    −70…+200 °C
  • Beskrivelse:
    Spring-loaded V4A tube Ø 3 mm with slim insertion tip, with plastic handle, anti-buckling glanding,1 m highly flexible silicone cable and 3,5 mm gold plated jack connector
    Response time (T90): water 0,4 m/s < 2 s, air 2 m/s ~40 s
  • Seq:
    01