Counting chambers, Bürker
Cell counting chamber, bright lined, with clamps, Bürker, 0,10 mm
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
: DIN 12847 and the German Calibration Ordinance.
: Supplied in a transparent box.
Specifikationer
- Kammerdybde:0,10 mm
- Beskrivelse:Cell counting chamber, bright lined, with clamps, Bürker
- Seq:04