Counting chambers, Bürker
Tællekamre, mørk kant, uden clips, Bürker, 0,10 mm
Katalog # 631-0920
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
Certifications: DIN 12847 and the German Calibration Ordinance.
Delivery information: Supplied in a transparent box.
Specifikationer
- Kammerdybde:0,10 mm
- Beskrivelse:Tællekamre, mørk kant, uden clips, Bürker
- Seq:06