Propper, diamantslib, DURAN®
24/29, Med spids bund
Katalog # 217-0792
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Pointed
- Diamond-ground
Specifikationer
- NS:24/29
- Model:Med spids bund