Orders LinkContactUsLinkComponent
Propper, diamantslib, DURAN®
19/26, Med spids bund
  217-0791
Propper, diamantslib, DURAN®
  217-0791
Propper, diamantslib, DURAN®
Propper, diamantslib, DURAN®
19/26, Med spids bund
2.150,00 kr
1 * 100 Items
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Pointed
  • Diamond-ground
  • NS:
    19/26
  • Model:
    Med spids bund