Propper, diamantslib, DURAN®
34/35, Med spids bund
Catalog # 217-0794
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Pointed
- Diamond-ground
Specifications
- NS:34/35
- Model:Med spids bund