Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Spherical ground joints, cup, DURAN®

Spherical ground joints, cup, DURAN®

S35/20, 1,8 mm, 24 mm
Katalog # SCOT286325403
Leverandør:   DWK Life Sciences
Spherical ground joints, cup, DURAN®
Katalog # SCOT286325403
Leverandør:  DWK Life Sciences
Spherical ground joints, cup, DURAN®
Spherical ground joints, cup, DURAN®

S35/20, 1,8 mm, 24 mm

Price
1.480,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Inches size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Through use of flexible spherical joint connections, apparatus can be assembled free from stresses.

*non-DIN size

Certifications: DIN 12244, DIN 12264

Specifikationer
  • Top:
    S35/20
  • Rør udv. Ø:
    24 mm
  • Vægtykkelse:
    1,8 mm
  • Seq:
    10
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.