Orders LinkContactUsLinkComponent
Adapters, expansion, DURAN®
Expansion, 14/23 NS, 19/26 NS
Katalog # SCOT286535906
Leverandør:   DWK Life Sciences
Adapters, expansion, DURAN®
Katalog # SCOT286535906
Leverandør:  DWK Life Sciences
Adapters, expansion, DURAN®
Adapters, expansion, DURAN®
Expansion, 14/23 NS, 19/26 NS
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
DURAN®, borosilicate glass 3,3 clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Specifikationer
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    19/26 NS
  • Model:
    Expansion
  • Seq:
    01