Adapters, expansion, DURAN®
Expansion, 14/23 NS, 19/26 NS
Katalog # SCOT286535906
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3,3 clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Specifikationer
- Konusstr.:14/23 NS
- Kappestr.:19/26 NS
- Model:Expansion
- Seq:01